Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv8.1 Antibody, Novus Biologicals™
SDP

Catalog No. p-7110700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237847 100 μL
NBP23784720 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP237847 Supplier Novus Biologicals Supplier No. NBP237847
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kv8.1 Polyclonal specifically detects Kv8.1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Kv8.1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q6PIU1
Gene Alias HNKA, KCNB3, KV2.3, KV8.1, Neuronal potassium channel alpha subunit HNKA, potassium channel Kv8.1, potassium channel, subfamily V, member 1, potassium voltage-gated channel subfamily V member 1, Voltage-gated potassium channel subunit Kv8.1
Gene Symbols KCNV1
Host Species Rabbit
Immunogen The immunogen for Anti-KCNV1 antibody is: synthetic peptide directed towards the N-terminal region of Human KCNV1.. Peptide sequence: LLDSPLDSGSLTSLDSSVFCSEGEGEPLALGDCFTVNVGGSRFVLSQQAL
Molecular Weight of Antigen 55 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27012
Target Species Human, Mouse, Rat, Pig, Canine, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.