Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv8.2 Antibody, Novus Biologicals™
SDP

Catalog No. NB176682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176682 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176682 Supplier Novus Biologicals Supplier No. NBP312995
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Kv8.2 Polyclonal antibody specifically detects Kv8.2 in Human samples. It is validated for Western Blot

Specifications

Antigen Kv8.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias KV11.1, Kv8.2, MGC120515, potassium channel, subfamily V, member 2, potassium voltage-gated channel subfamily V member 2, RCD3B, voltage-gated potassium channel Kv8.2, Voltage-gated potassium channel subunit Kv8.2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Kv8.2. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 169522
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.