Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv9.3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305922JF549
Description
Kv9.3 Polyclonal antibody specifically detects Kv9.3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Kv9.3 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
delayed-rectifier K(+) channel alpha subunit 3, KV9.3, MGC9481, potassium voltage-gated channel subfamily S member 3, potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3, Shab-related delayed-rectifier K+ channel alpha subunit 3, voltage-gated potassium channel protein Kv9.3, voltage-gated potassium channel subunit Kv9.3 | |
Produced in rabbits immunized with E. coli-derived Human Kv9.3 fragment. | |
0.1 mL | |
Neuroscience | |
3790 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction