Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KVbeta1 (KCNAB1) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560743 Supplier Invitrogen™ Supplier No. PA560743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSQISEENTK LRRQSGFSVA GKDKSPKKAS ENAKDSSLSP SGESQLRARQ LALLREVEMN WYLKLCDLSS EHTTVCTTGM Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 81%.

Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNAB1 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KVbeta1 (KCNAB1)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNAB1
Gene Accession No. Q14722
Gene Alias AKR6A3; Akr8a8; hKvb3; hKvBeta3; K(+) channel subunit beta-1; K+ channel Beta1a chain; KCNA1B; KCNAB1; Kvb1; Kvb1.3; KVBETA1; Kv-beta-1; Kvbeta1.1; KvBeta3; mKv(beta)1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel subunit Kv beta 1; potassium channel subunit Kv beta 1.1; potassium channel subunit Kv beta 1.2; potassium channel subunit Kv beta 1.3; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium channel, voltage-gated shaker related subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage-gated channel shaker-related subfamily beta member 1; potassium voltage-gated channel subfamily A member regulatory beta subunit 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; voltage-gated potassium channel subunit beta-1
Gene Symbols KCNAB1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human KCNAB1 (Product #RP-95686)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.