Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KVbeta2 (KCNAB2) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568936 Supplier Invitrogen™ Supplier No. PA568936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 100%.

Kcnab2 is a voltage-gated potassium channels (KV) belong to the 6-TM family of potassium channel that also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). Kcnab2 is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. Alternative splicing of the Kcnab2 gene results in multiple transcript variants encoding distinct isoforms. Diseases associated with KCNAB2 include Chromosome 1P36 Deletion Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KVbeta2 (KCNAB2)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNAB2
Gene Accession No. P62482, Q13303
Gene Alias AKR6A5; Ckbeta2; F5; HKvbeta2; HKvbeta2.1; HKvbeta2.2; I2rf5; K(+) channel subunit beta-2; KCNA2B; KCNAB2; KCNAB2.1; KCNAB2.2; Kcnb3; KCNK2; KVB2; Kvbeta2; Kv-beta-2; Kvbeta2.1; Kvbeta2.2; neuroimmune protein F5; potassium channel; potassium channel Kv beta 2.1 subunit; potassium channel Kv beta 2.2 subunit; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium channel, voltage-gated shaker-related subfamily A regulatory beta subunit 2; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage-gated channel subfamily A regulatory beta subunit 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2, transcript variant 1; potassium voltage-gated channel, shaker-related subfamily, beta member 2, transcript variant 2; voltage-gated potassium channel subunit beta-2
Gene Symbols KCNAB2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KCNAB2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16498, 8514
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.