Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KvBeta3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | KvBeta3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
KvBeta3 Polyclonal specifically detects KvBeta3 in Mouse samples. It is validated for Western Blot.Specifications
KvBeta3 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
9196 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
AKR6A9, K(+) channel subunit beta-3, KCNA3.1B, KCNA3BK+ channel beta-3 subunit, Kv-beta-3, MGC116886, potassium channel, voltage-dependent, beta-3 subunit, potassium voltage-gated channel, shaker-related subfamily, beta member 3, voltage-gated potassium channel subunit beta-3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 285-404 of human KvBeta3 (NP_004723.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title