Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KW 3902
GREENER_CHOICE

Catalog No. 416710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-671-0 10 mg
41-675-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-671-0 Supplier Tocris Bioscience™ Supplier No. 4167/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective A1 antagonist

Selective adenosine A1 receptor antagonist; displays 890-fold selectivity for rat A1 receptors over A2A receptors (Ki values are 0.19 and 170 nM respectively). Displays no effect on recombinant rat A3 receptors expressed on CHO cells at concentrations up to 10 μM. Exhibits diuretic and renal protective effects in rats.

Chemical Identifiers

CAS 136199-02-5
Molecular Formula C20H28N4O2

Specifications

Quantity 10 mg
Formula Weight 356.46
Purity >99%
Molecular Weight (g/mol) 356.47
Chemical Name or Material KW 3902
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.