Learn More
Invitrogen™ KXD1 Monoclonal Antibody (4C9)

Description
Peptide Sequence: FRRIRTLKGK LARQHPEAFS HIPEASFLEE EDEDPIPPST TTTIATSEQS TGSCDTSPDT VSPSLSPGFE DLSHVQAGSP AINGRSQTDD EEMTGE.
Specifications
Specifications
| Antigen | KXD1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | KXD1 |
| Gene Accession No. | Q9BQD3 |
| Gene Alias | BORCS4; C19orf50; KXD1; KXDL; KxDL motif containing 1; kxDL motif-containing protein 1; MST096; MSTP096; UPF0459 protein C19orf50 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.