Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KXD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559602 Supplier Invitrogen™ Supplier No. PA559602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARQHPEAFSH IPEASFLEEE DEDPIPPSTT TTIATSEQST GSCDTSPDTV SPSLSPGFED LSHVQAGSPA INGRSQTDDE EMTGE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 85%.

The function of the C19orf50 protein has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KXD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KXD1
Gene Accession No. Q9BQD3
Gene Alias BORCS4; C19orf50; KXD1; KXDL; KxDL motif containing 1; kxDL motif-containing protein 1; MST096; MSTP096; UPF0459 protein C19orf50
Gene Symbols KXD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KXD1. Recombinant protein control fragment (Product #RP-99002).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.