Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KY 02111
GREENER_CHOICESDP

Catalog No. 47315 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-315 5 mg
47-312-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-315 Supplier Tocris Bioscience™ Supplier No. 4731/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibits canonical Wnt signaling. Promotes differentiation of human ESCs and iPSCs into cardiomyocytes

KY 02111 promotes differentiation of human ESCs and iPSCs into cardiomyocytes when used after day three in culture; differentiation efficiency increases when used after initial culture with CHIR 99021 (Cat. No. 4423). Induces downregulation of Wnt signaling target genes; inhibits canonical Wnt signaling in a manner distinct from other known Wnt inhibitors.

Specifications

Chemical Name or Material N-(6-Chloro-2-benzothiazolyl)-3,4-dimethoxybenzenepropanamide
CAS 1118807-13-8
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C18H17ClN2O3S
Formula Weight Observed MW: 376.86
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.