Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597916
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597916 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597916 Supplier Invitrogen™ Supplier No. PA597916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KY Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Probable cytoskeleton-associated protease required for normal muscle growth. Involved in function, maturation and stabilization of the neuromuscular junction. May act by cleaving muscle-specific proteins such as FLNC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KY
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KY
Gene Accession No. Q8NBH2
Gene Alias CD1; D9Mgc44e; ky; Kyphoscoliosis peptidase
Gene Symbols KY
Host Species Rabbit
Immunogen Recombinant Human Kyphoscoliosis peptidase protein (23-170AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339855
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.