Learn More
Invitrogen™ KYNU Recombinant Rabbit Monoclonal Antibody (004)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529723
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse KYNU.
Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
KYNU | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9CXF0 | |
Kynu | |
Recombinant Mouse KYNU protein (Met1-Ser464). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
004 | |
Unconjugated | |
Kynu | |
4432411A05Rik; HAMAP-Rule:MF_03017}; KYNU; kynureninase; kynureninase (L-kynurenine hydrolase); kynureninase {ECO:0000255; kynureninase-like protein; KYNUU; L-kynurenine hydrolase; L-kynurenine hydrolase {ECO:0000255 | |
Rabbit | |
Protein A | |
RUO | |
70789 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.