Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kynureninase Antibody (005), Biotin

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290526B
Description
Kynureninase Monoclonal specifically detects Kynureninase in Mouse samples. It is validated for ELISA.Specifications
Kynureninase | |
Monoclonal | |
Biotin | |
EC 3.7.1.3, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Kynureninase (Q9CXF0; Met1-Ser464). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
5 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
8942 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction