Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KYP 2047
GREENER_CHOICESDP

Catalog No. 62725 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-725 5 mg
62-722-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-725 Supplier Tocris Bioscience™ Supplier No. 6272/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity prolyl oligopeptidase inhibitor (Ki = 0.023 nM); anti-angiogenic, neuroprotective and brain penetrant

KYP 2047 is a high affinity prolyl oligopeptidase (POP) inhibitor (Ki = 0.023 nM). Clears α-synuclein aggregates induced by oxidative stress in neuronal cells. Inhibits the formation of AcSDKP from its precursor 43-mer thymosin β4 (Tβ4). Induces angiogenesis via POP inhibition. It reduces tau aggregation in tau-transfected HEK-293 cells and N2A cells as well as in human iPSC-derived neurons from patients with frontotemporal dementia. In vivo, treatment with KYP 2047 of transgenic mice with tauopathy, reduces tau aggregation in the brain and cerebrospinal fluid and slows cognitive decline. Anti-angiogenic and neuroprotective. Brain penetrant.

Specifications

Chemical Name or Material (2S)-1-[[(2S)-1-(1-Oxo-4-phenylbutyl)-2-pyrrolidinyl]carbonyl]-2-pyrrolidinecarbonitrile
CAS 796874-99-2
Quantity 5 mg
Molecular Formula C20H25N3O2
Formula Weight 339.44
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.