Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-168,049
GREENER_CHOICESDP

Catalog No. 231150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-115-0 50 mg
23-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-115-0 Supplier Tocris Bioscience™ Supplier No. 2311/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, orally active human glucagon receptor antagonist

Very potent and selective, non-competitive antagonist of the human glucagon receptor (hGR). Binds with high affinity to human GR (IC50 = 3.7 nM), and moderate affinity to murine and canine GRs (IC50 values are 63 and 60 nM respectively). In contrast, displays poor affinity for rat, guinea pig, and rabbit glucagon receptors (IC50 > 1 μM). In functional studies, inhibits glucagon-stimulated cAMP synthesis in CHO cells expressing hGR (IC50 = 41 nM), and in murine liver membranes. Orally active in vivo.

Chemical Identifiers

CAS 191034-25-0
Molecular Formula C24H20BrClN2O

Specifications

Quantity 50 mg
Formula Weight 467.79
Purity >99%
Molecular Weight (g/mol) 467.791
Chemical Name or Material L-168, 049
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.