Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-2-Aminoadipic acid, 98%

Catalog No. AAL1392403
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAL1392403

Supplier: Thermo Scientific Chemicals L1392403

Only null left
Add to Cart

CAS: 1118-90-7 | C6H11NO4 | 161.16 g/mol

L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in 1N HCl (50 mg/ml) and water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-90-7
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00002636
InChI Key OYIFNHCXNCRBQI-BYPYZUCNSA-N
Synonym l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid
PubChem CID 92136
ChEBI CHEBI:37023
IUPAC Name (2S)-2-aminohexanedioic acid
SMILES N[C@@H](CCCC(O)=O)C(O)=O

Specifications

Melting Point ∽206°C (decomposition)
Quantity 1 g
Beilstein 1724348
Solubility Information Soluble in 1N HCl (50mg/ml) and water (slightly).
Optical Rotation +25° (c=2 in 5N HCl)
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material L-2-Aminoadipic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.