Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ L-365,260
GREENER_CHOICESDP

Catalog No. 276750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-675-0 50 mg
27-671-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-675-0 Supplier R&D Systems™ Supplier No. 2767/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective CCK2 antagonist

Selective cholecystokinin receptor 2 (CCK2) antagonist (IC50 values are 2 and 280 nM at CCK2 and CCK1 receptors respectively) that is inactive at a range of other receptors including opiate, muscarinic acetylcholine, α- and β adrenergic, histamine, angiotensin and bradykinin receptors.

Chemical Identifiers

CAS 118101-09-0
Molecular Formula C24H22N4O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 398.46
Purity >99%
Molecular Weight (g/mol) 398.466
Chemical Name or Material L-365, 260
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.