Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-368,899 hydrochloride
GREENER_CHOICE

Catalog No. 26411
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-411 1 mg
26-411-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-411 Supplier Tocris Bioscience™ Supplier No. 2641/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, non-peptide oxytocin receptor antagonist

Potent, non-peptide and orally active oxytocin receptor antagonist (IC50 = 8.9 nM) that displays > 40-fold selectivity over vasopressin V1a and V2 receptors (IC50 values are 370 and 570 nM respectively). Antagonizes oxytocin-induced uterine contractions in vitro and in vivo.

Chemical Identifiers

CAS 160312-62-9
Molecular Formula C26H43ClN4O5S2

Specifications

Quantity 1 mg
Formula Weight 591.23
Purity >97%
Molecular Weight (g/mol) 591.223
Chemical Name or Material L-368, 899 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.