Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propionic acid, sodium salt, 99.0-100.5%
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 285°C to 286°C |
Color | White |
Flash Point | >250°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.0-100.5% |
Packaging | Plastic drum |
Linear Formula | C2H5CO2Na |
Quantity | 5 kg |
Beilstein | 02, 234 |
Merck Index | 15, 8798 |
Show More |
Safety and Handling
Hazard Statement | GHS H Statement: Causes skin irritation. Harmful in contact with skin. |
Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge |
EINECSNumber | 205-290-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction