Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-651,582
GREENER_CHOICESDP

Catalog No. 308610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-861-0 10 mg
30-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-861-0 Supplier Tocris Bioscience™ Supplier No. 3086/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ca2+ channel blocker

Orally active Ca2+ channel blocker; inhibits K+- and carbachol-stimulated Ca2+ influx (IC50 values are 500 and 935 nM respectively). Exhibits antiproliferative, antiangiogenic and antimetastatic activity in vivo and displays selectivity towards numerous mismatch repair-deficient tumor cell lines in vitro.

Chemical Identifiers

CAS 99519-84-3
Molecular Formula C17H12Cl3N5O2

Specifications

Quantity 10 mg
Formula Weight 424.67
Purity >98%
Molecular Weight (g/mol) 424.67
Chemical Name or Material L-651, 582
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.