Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ L-651,582
GREENER_CHOICESDP

Catalog No. 308650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-865-0 50 mg
30-861-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-865-0 Supplier R&D Systems™ Supplier No. 3086/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Ca2+ channel blocker

Orally active Ca2+ channel blocker; inhibits K+- and carbachol-stimulated Ca2+ influx (IC50 values are 500 and 935 nM respectively). Exhibits antiproliferative, antiangiogenic and antimetastatic activity in vivo and displays selectivity towards numerous mismatch repair-deficient tumor cell lines in vitro.

Chemical Identifiers

CAS 99519-84-3
Molecular Formula C17H12Cl3N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 424.67
Purity >98%
Molecular Weight (g/mol) 424.67
Chemical Name or Material L-651, 582
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.