Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-689,560
GREENER_CHOICE

Catalog No. 074210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-421-0 10 mg
07-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-421-0 Supplier Tocris Bioscience™ Supplier No. 0742/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent NMDA antagonist

L-689,560 is a very potent antagonist at the glycine-NMDA site.

Specifications

Chemical Name or Material trans-2-Carboxy-5,7-dichloro-4-phenylaminocarbonylamino-1,2,3,4-tetrahydroquinoline
CAS 139051-78-8
Assay Percent Range >99%
Quantity 10 mg
Solubility Soluble to 25mM in DMSO
Target NMDA Receptor Antagonists
Molecular Formula C17H15Cl2N3O3
Formula Weight Observed MW: 380.23
Purity 99%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.