Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-692,585, Tocris Bioscience™
SDP

Catalog No. 226110 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-611-0 Supplier R&D Systems Supplier No. 2261/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent, non-peptide ghrelin receptor agonist

Highly potent, non-peptide ghrelin receptor (GHS-R1a) agonist (Ki = 0.8 nM). Increases plasma GH levels in vivo and is 2- to 2.5-fold more potent than GHRP-6.

Chemical Identifiers

CAS 145455-35-2
Molecular Formula C32H37N7O3
Molecular Weight (g/mol) 567.694
InChI Key AYBCFPXLXOLPIZ-JIPXPUAJSA-N
Synonym chembl13600, 3-2r-2-hydroxypropyl amino-3-methyl-n-3r-2,3,4,5-tetrahydro-2-oxo-1-2'-1h-tetrazol-5-yl 1,1'-biphenyl-4-yl methyl-1h-1-benzazepin-3-yl-butanamide, 3-2r-2-hydroxypropyl amino-3-methyl-n-3r-2-oxo-1-2'-1h-1,2,3,4-tetrazol-5-yl-1,1'-biphenyl-4-yl methyl-4,5-dihydro-3h-1-benzazepin-3-yl butanamide
PubChem CID 9872740
IUPAC Name 3-[[(2R)-2-hydroxypropyl]amino]-3-methyl-N-[(3R)-2-oxo-1-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]-4,5-dihydro-3H-1-benzazepin-3-yl]butanamide
SMILES CC(CNC(C)(C)CC(=O)NC1CCC2=CC=CC=C2N(C1=O)CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5)O

Specifications

Quantity 10 mg
Formula Weight 567.69
Percent Purity >98%
Chemical Name or Material L-692, 585
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.