Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-755,507

Catalog No. 219750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-975-0 50 mg
21-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-975-0 Supplier Tocris Bioscience™ Supplier No. 2197/50
This item is not returnable. View return policy

Very potent and selective β3 partial agonist

L-755,507 is a potent β3-adrenergic receptor partial agonist > 1000-fold selective over β1- and β2-adrenoceptors (EC50 values are 0.43, 580 and > 10000 nM for activation of cloned human β3-, β1- and β2-adrenoceptors respectively). Stimulates lipolysis in rhesus adipocytes in vitro (EC50 = 3.9 nM). Enhances CRISPR-mediated homology-directed repair (HDR) efficiency 2-3-fold for large fragments and ∽9-fold for point mutations, in human induced pluripotent stem cells (iPSCs).

Specifications

Quantity 50 mg
Product Type L-755,507
Purity >98%
Molecular Weight (g/mol) 584.73
Inhibitors L-755,507

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.