Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine 2-ethylbutyl ester hydrochloride, Thermo Scientific™

Catalog No. p-7227421
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Alpha Vector ALANINEETHYLBUTYLESTERHYDROCHLORIDE
Chemical Name or Material L-Alanine 2-ethylbutyl ester hydrochloride
CAS 946511-97-3
Melting Point 75°C to 80°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C9 H19 N O2 . H Cl
Quantity 1 g
MDL Number 32660031
Synonym (S)-2-Ethylbutyl 2-aminopropanoate hydrochloride
Molecular Weight (g/mol) 209.74
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.