Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine, 99%, specified according to the requirements of Ph.Eur.

Catalog No. p-4839223
Click to view available options
Quantity:
250 g
1 kg
2.5 kg

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Melting Point 314.0°C to 316.0°C
Assay Percent Range 99%
Linear Formula CH3CH(NH2)CO2H
Specific Rotation + 14.50
Solubility Information Solubility in water: 166.5g/l in water (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents
Quantity 250 g
Formula Weight 89.09
Percent Purity 99%
Chemical Name or Material L-Alanine, specified according to the requirements of Ph.Eur.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.