Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine Ethyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A249425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A249425G 25 g
A24945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A249425G Supplier TCI America Supplier No. A249425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1115-59-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00063662
InChI Key JCXLZWMDXJFOOI-UHFFFAOYNA-N
Synonym l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride
PubChem CID 2724356
IUPAC Name hydrogen ethyl 2-aminopropanoate chloride
SMILES [H+].[Cl-].CCOC(=O)C(C)N

Specifications

Melting Point 79°C
Color White
Quantity 25 g
Formula Weight 153.61
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine Ethyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.