Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Alanine hydrochloride Solution 100 mM amino acid in 0.1 M HCl, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
5 mL
Description
Featured Industries: Food and Beverages

Specifications
Specifications
CAS | 6003-05-0 |
Concentration | 100 mM amino acid in 0.1 M HCl |
Linear Formula | C3H7 NO2 · HCl |
MDL Number | MFCD03791086 |
Type | Analytical Standard |
Quantity | 5 mL |
Formula Weight | 125.55 |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction