Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Alanine methyl ester hydrochloride, 99%

Manufacturer:  Alfa Aesar™ A1017806

Quantity 5g

 View more versions of this product

Catalog No. AAA1017806


  Check Availability
Add to cart

Specifications

Description

Applications
L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.

Solubility
Soluble in Water (100 mg/ml).

Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

2491-20-5
139.58
3594033
L-Alanine methyl ester hydrochloride
C4H9NO2·HCl
MFCD00063663
H-Ala-OMe.HCl

Specifications

2491-20-5
139.58
3594033
106° to 110°C
+6.5° (c=10 in methanol)
99%
C4H9NO2·HCl
MFCD00063663
H-Ala-OMe.HCl
5g
L-Alanine methyl ester hydrochloride

Safety and Handling

SDS & Safety Documents

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit