Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine tert-butyl ester hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13404-22-3
C7H16ClNO2
181.66
MFCD00035524
WIQIWPPQGWGVHD-JEDNCBNOSA-N
h-ala-otbu.hcl,tert-butyl l-alaninate hydrochloride,l-alanine tert-butyl ester hydrochloride,s-tert-butyl 2-aminopropanoate hydrochloride,tert-butyl alaninate hydrochloride,h-ala-otbu inverted exclamation mark currencyhcl,tert-butyl 2s-2-aminopropanoate hydrochloride,l-alanine t-butyl ester hydrochloride,alanine, 1,1-dimethylethyl ester, hydrochloride,l-alanine, 1,1-dimethylethyl ester, hydrochloride
11275390
tert-butyl (2S)-2-aminopropanoate;hydrochloride
CC(C(=O)OC(C)(C)C)N.Cl
13404-22-3
C7H16ClNO2
181.66
MFCD00035524
WIQIWPPQGWGVHD-JEDNCBNOSA-N
h-ala-otbu.hcl,tert-butyl l-alaninate hydrochloride,l-alanine tert-butyl ester hydrochloride,s-tert-butyl 2-aminopropanoate hydrochloride,tert-butyl alaninate hydrochloride,h-ala-otbu inverted exclamation mark currencyhcl,tert-butyl 2s-2-aminopropanoate hydrochloride,l-alanine t-butyl ester hydrochloride,alanine, 1,1-dimethylethyl ester, hydrochloride,l-alanine, 1,1-dimethylethyl ester, hydrochloride
11275390
tert-butyl (2S)-2-aminopropanoate;hydrochloride
CC(C(=O)OC(C)(C)C)N.Cl
Packaging Glass bottle
Formula Weight 181.66
Physical Form Solid or Crystalline Powder
Color White
Quantity 5g
Assay 99%

EINECSNumber : 236-497-8