Learn More
Description
Available as part of the Group III mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . DL Mixture and D-isomer also available.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 183.1 |
| Purity | >99% |
| Molecular Weight (g/mol) | 183.1 |
| Chemical Name or Material | L-AP4 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

