Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18611195 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-195 100 g Amber Glass Bottle
18-611-197 25 g Amber Glass Bottle
18-611-194 1 kg Amber Glass Bottle
18-611-196 12 kg Amber Glass Bottle
18-611-198 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-195 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AR120100GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74-79-3

Specifications

CAS Max % 1
Heavy Metals 20 ppm
Lead (Pb) 10 ppm
Loss on Drying 0.01
Packaging Amber Glass Bottle
Residue after Ignition 0.002
Specific Rotation +26.0° to +27.7°
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade Reagent
Chemical Name or Material L-Arginine
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.