Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine, Free Base, cGMP, USP, EP, JP, bioCERTIFIED™, 2.5 kg, Spectrum Chemical
SDP

Catalog No. 18615431
Click to view available options
Quantity:
2.5 kg

100% Tested. 100% Transparent. 100% Trusted.

Whether it's upstream bioprocessing or downstream, Spectrum Chemical's bioCERTIFIED™ products are part of a comprehensive quality management system that includes APIs, excipients and other chemicals that carry additional certifications and documentation.

Each bioCERTIFIED™ chemical undergoes expanded testing for bioburden, endotoxin, elemental impurities and other tests required by the monographs. Products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Key attributes of Spectrum Chemicals bioCERTIFIED™ portfolio include:

  • Complete lot analysis including tests for endotoxin, bioburden and elemental impurities
  • Comprehensive regulatory and scientific documentation
  • Change control, batch traceability and supply chain transparency
  • Extensive product breadth and Packaging flexibility

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 98.5 to 101.5%
Grade cGMP/USP/EP/JP/bioCERTIFIED
Chemical Name or Material L-Arginine, Free Base
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.