Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Arginine, Free Base, Fisher BioReagents
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Description
L-Arginine is an essential amino acid used in biochemical research applications.

Specifications
Specifications
Melting Point | 223°C |
pH | 11.4 |
Color | White |
Identification | Pass Test |
Infrared Spectrum | Conforms to reference |
Assay Percent Range | ≥99 % |
Loss on Drying | 0.3% max. |
Packaging | Poly Bottle |
Specific Rotation | +26.3 to +27.7° (+ or -) |
Merck Index | 15, 770 |
Show More |
Safety and Handling

Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction