Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ L-Ascorbic acid
GREENER_CHOICESDP

Catalog No. RD10000120 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000120 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000120 Supplier R&D Systems™ Supplier No. TB4055RMU/500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. L-Ascorbic acid synthesized to Ancillary Material Grade

TB4055-RMU is an Ancillary Material Grade version of product L-Ascorbic acid (Cat. No. 4055). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.

Specifications

Chemical Name or Material 3-Oxo-L-gulofuranolactone
CAS 50-81-7
Quantity 500 mg
Product Line Tocris
Target iPSC Generation
Molecular Formula C6H8O6
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.