Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate 98.0+%, TCI America™
SDP

Catalog No. A252125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A252125G 25 g
A25215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A252125G Supplier TCI America Supplier No. A252125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 113170-55-1
Molecular Formula C12H18Mg3O18P2
Molecular Weight (g/mol) 585.12
MDL Number MFCD08063372
InChI Key IRBRZNUQJUHOBT-UHFFFAOYNA-N
PubChem CID 129894323
IUPAC Name bis({[5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-yl]oxy}phosphonic acid) trimagnesium
SMILES [Mg].[Mg].[Mg].OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O.OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O

Specifications

Quantity 25 g
Formula Weight 579.07
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate
EINECSNumber (5)-5982
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.