Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-Ascorbic acid
GREENER_CHOICESDP

Catalog No. 405550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-555-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-555-0 Supplier Tocris Bioscience™ Supplier No. 4055/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enhances the generation of iPSCs; increases reprogramming efficiency

Inhibitor of Cav3.2 channels (IC50 = 6.5 μM); displays no effect on Cav3.1 or Cav3.3 channels heterologously expressed in HEK 293 cells. Also enhances the generation of induced pluripotent stem cells (iPSCs) from mouse and human somatic cells by increasing reprogramming efficiency. Naturally occurring antioxidant.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6

Specifications

Quantity 50 mg
Formula Weight 176.12
Purity >99%
Molecular Weight (g/mol) 176.12
Chemical Name or Material L-Ascorbic acid
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.