Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Asparaginase Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178086AF405
Description
L-Asparaginase Polyclonal antibody specifically detects L-Asparaginase in E. coli samples. It is validated for Western Blot, ELISASpecifications
L-Asparaginase | |
Polyclonal | |
Western Blot, ELISA | |
AnsA, AnsB, Asparaginase, Colaspase, Cytoplasmic asparaginase I, L ASNase I, L ASNase II, L asparaginase 1, L asparaginase 2, L asparaginase I, L asparaginase II, L asparaginase II precursor, L asparagine amidohydrolase I, L asparagine amidohydrolase II | |
L-L-Asparaginase [E. coli] (Uniprot: P00805) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. |
Western Blot, ELISA | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Multi-step | |
RUO | |
E. coli | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction