Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, ACS reagent

Catalog No. AC449060010
Click to view available options
Quantity:
250 g
1 kg

CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO

Specifications

Melting Point 27.0°C to 30.0°C
Density 1.0955g/mL
Boiling Point 268.0°C
Flash Point 179°C
Refractive Index 1.4750 to 1.4790
Linear Formula (HOCH2CH2)2NH
Quantity 1 kg
Merck Index 15,3118
Specific Gravity 1.0955
Solubility Information Solubility in water: 954g/L (20°C).
Viscosity 352 mPa.s (30°C)
Formula Weight 105.14
Percent Purity ≥98.5% (GC) 1% max. (2-Aminoethanol) (GC) 1.0% max. (Triethanolamine) (GC)
Grade ACS Reagent
Chemical Name or Material Diethanolamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 203-868-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.