Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic acid diethyl ester hydrochloride, 98%

Catalog No. p-7055401
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 16115-68-7 | C8H16ClNO4 | 225.669 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16115-68-7
Molecular Formula C8H16ClNO4
Molecular Weight (g/mol) 225.669
MDL Number MFCD00038910
InChI Key AJOXZAAREAYBQR-RGMNGODLSA-N
Synonym l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
PubChem CID 12416466
IUPAC Name diethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CCOC(=O)CC(C(=O)OCC)N.Cl

Specifications

Quantity 5 g
Formula Weight 225.67
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Aspartic acid diethyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.