Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-beta-Proline, 98+%

Catalog No. aah57895md
Click to view available options
Quantity:
250 mg
1 g

CAS: 72580-53-1 | C5H9NO2 | 115.132 g/mol

It is an agrochemical and pharmacetical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical and pharmacetical intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72580-53-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD06738463
InChI Key JAEIBKXSIXOLOL-BYPYZUCNSA-N
Synonym s-pyrrolidine-3-carboxylic acid, s-+-pyrrolidine-3-carboxylic acid, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3s-pyrrolidine-3-carboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-, a-proline, pubchem11126
PubChem CID 1501969
IUPAC Name (3S)-pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O

Specifications

Melting Point 185°C to 191°C
Beilstein 3537293
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 115.13
Optical Rotation +18° (c=2 in Water)
Percent Purity ≥98%
Chemical Name or Material L-beta-Proline
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.