Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(-)-Bornyl acetate, 95%

Catalog No. AAB2295736
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

Specifications

Density 0.98
Boiling Point 223°C to 224°C
Flash Point 84°C (184°F)
Refractive Index 1.463
Quantity 500 g
Beilstein 2502036
Merck Index 14,1339
IUPAC Name (1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
Formula Weight 196.29
Percent Purity 95%
Chemical Name or Material L-(-)-Bornyl acetate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 227-101-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.