Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Bornyl acetate, 95%

Catalog No. AC106551000
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg

CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
IUPAC Name [(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

Specifications

Color Colorless
Density 0.9875g/mL
Boiling Point 223°C to 224°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (as C12H20O2) Total esters (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.465
Quantity 100 g
Beilstein 06,82
Merck Index 15,1341
Specific Gravity 0.9875
Specific Rotation -38
Solubility Information Solubility in water: .. Other solubilities: miscible with 95% alcohol and ether
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material L-Bornyl acetate, 95%
Show More Show Less
EINECSNumber 227-101-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.