Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Carnitine, 98+%

Catalog No. p-7023752
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 541-15-1 | C7H15NO3 | 161.201 g/mol

L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

Solubility
Soluble in water, pyridine, dimethyl sulfoxide, methanol and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-15-1
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.201
MDL Number MFCD00038747
InChI Key PHIQHXFUZVPYII-ZCFIWIBFSA-N
Synonym l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
PubChem CID 10917
ChEBI CHEBI:16347
IUPAC Name (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)CC(CC(=O)[O-])O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 10 g
Beilstein 4292315
Sensitivity Hygroscopic
Merck Index 14,1849
Solubility Information Soluble in water,pyridine,dimethyl sulfoxide,methanol and ethanol.
Formula Weight 161.2
Percent Purity ≥99%
Chemical Name or Material L-Carnitine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-768-0
RTECSNumber BP2980000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.