Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Carnitine-(trimethyl-d9) inner salt, Analytical Standard, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7193437 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg

Specifications

CAS 126827-79-0
Linear Formula C7D9H6NO3
Synonym (R)-3-Carboxy-2-hydroxy-N,N,N-tri(methyl-d3)-1-propanaminium inner salt
Quantity 10 mg
Formula Weight 170.25
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.