Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metaborate tetrahydrate, 98.5%, extra pure

Catalog No. AC211630025
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 10555-76-7 | BH8NaO6 | 137.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10555-76-7
Molecular Formula BH8NaO6
Molecular Weight (g/mol) 137.86
MDL Number MFCD00149205
InChI Key JAKYJVJWXKRTSJ-UHFFFAOYSA-N
Synonym sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
PubChem CID 23694267
SMILES O.O.O.O.[Na+].[O-]B=O

Specifications

Melting Point 57.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Assay Percent Range 98.5%
Linear Formula NaBO2·4H2O
Merck Index 10,8477
Solubility Information Solubility in water: 41.9% (20°C)
IUPAC Name sodium;oxido(oxo)borane;tetrahydrate
Formula Weight 137.86
Percent Purity 98.5%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium metaborate tetrahydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.