Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis)

Catalog No. AAJ1403522
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Plastic Bottle

CAS: 7048-04-6 | C3H10ClNO3S | 175.63 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7048-04-6
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
PubChem CID 23462
IUPAC Name (2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Melting Point ∼175°C (decomposition)
Packaging Plastic Bottle
Linear Formula HSCH2CH(NH2)COOH.HCl.H2O
Beilstein 5158059
Quantity 100 g
Formula Weight 175.63
Percent Purity 98.5 to 101.5%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Cysteine Hydrochloride Monohydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-157-7
RTECSNumber HA2285000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.