Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Cysteine Methyl Ester Hydrochloride 99.0+%, TCI America™

Click to view available options
Quantity:
5 g
Specifications
Specifications
Color | White |
Quantity | 5 g |
Formula Weight | 171.64 |
Percent Purity | ≥99.0% (T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | L-Cysteine Methyl Ester Hydrochloride |
Safety and Handling
EINECSNumber | (9)-1594 |
RTECSNumber | HA2460000 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction