Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Cysteine Monohydrochloride, Monohydrate, FCC, 98-101.5%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical L-Cysteine Monohydrochloride, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Quantity | 100 g |
Percent Purity | 98 to 101.5% |
Grade | FCC |
Ignition Residue | 0.1% |
Loss on Drying | 8.0-12% |
Optical Rotation | +5.0° to +8.0° |
Packaging | Amber Glass Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction