Learn More
L-Cystine, Cell Culture Reagent
Description
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.
Solubility
It is soluble in water and HCL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
Specifications
Specifications
| Melting Point | >240°C (decomposition) |
| Color | White |
| Beilstein | 1728094 |
| Merck Index | 14,2782 |
| Solubility Information | It is soluble in water and HCL. |
| Density | 1.68 |
| Quantity | 100 g |
| Formula Weight | 240.3 |
| Optical Rotation | −219° (c=1 in 1N HCl) |
| Grade | Cell Culture Reagent |
| Show More |
Safety and Handling
| EINECSNumber | 200-296-3 |
| RTECSNumber | HA2690000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.